man

Test

CAS RN 127-41-3

CH3OH3CCH3CH3

CAS RN 127-41-3

IUPAC Name: (3E)-4-(2,6,6-trimethylcyclohex-2-en-1-yl)but-3-en-2-one
Synonyms: ionone α-ionone (natural) cyclocitrylideneacetone, α- α-cyclocitrylideneacetone
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: UZFLPKAIBPNNCA-BQYQJAHWNA-N
SMILES: CC(=O)\C=C\C1C(C)=CCCC1(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results