CAS RN 128-37-0
CAS RN 128-37-0
2,6-Di-tert-butyl-4-methylphenol, 99.8%, Thermo Scientific Chemicals
CAS: 128-37-0 Molecular Formula: C15H24O Molecular Weight (g/mol): 220.35 MDL Number: MFCD00011644 InChI Key: NLZUEZXRPGMBCV-UHFFFAOYSA-N Synonym: 2,6-di-tert-butyl-4-methylphenol,butylated hydroxytoluene,butylhydroxytoluene,2,6-di-tert-butyl-p-cresol,2,6-di-t-butyl-4-methylphenol,ionol,dbpc,dibunol,stavox,bht PubChem CID: 31404 ChEBI: CHEBI:34247 IUPAC Name: 2,6-ditert-butyl-4-methylphenol SMILES: CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
2,6-Di-tert-butyl-4-methylphenol, 99%, Thermo Scientific Chemicals
CAS: 128-37-0 Molecular Formula: C15H24O Molecular Weight (g/mol): 220.356 MDL Number: MFCD00011644 InChI Key: NLZUEZXRPGMBCV-UHFFFAOYSA-N Synonym: 2,6-di-tert-butyl-4-methylphenol,butylated hydroxytoluene,butylhydroxytoluene,2,6-di-tert-butyl-p-cresol,2,6-di-t-butyl-4-methylphenol,ionol,dbpc,dibunol,stavox,bht PubChem CID: 31404 ChEBI: CHEBI:34247 IUPAC Name: 2,6-ditert-butyl-4-methylphenol SMILES: CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
2,6-Di-tert-butyl-4-methylphenol, 99%, Thermo Scientific Chemicals
CAS: 128-37-0 Molecular Formula: C15H24O Molecular Weight (g/mol): 220.35 MDL Number: MFCD00011644 InChI Key: NLZUEZXRPGMBCV-UHFFFAOYSA-N Synonym: 2,6-di-tert-butyl-4-methylphenol,butylated hydroxytoluene,butylhydroxytoluene,2,6-di-tert-butyl-p-cresol,2,6-di-t-butyl-4-methylphenol,ionol,dbpc,dibunol,stavox,bht PubChem CID: 31404 ChEBI: CHEBI:34247 IUPAC Name: 2,6-ditert-butyl-4-methylphenol SMILES: CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
Butylated Hydroxytoluene, 99.5%, MP Biomedicals™
CAS: 128-37-0 Molecular Formula: C15H24O Molecular Weight (g/mol): 220.356 InChI Key: NLZUEZXRPGMBCV-UHFFFAOYSA-N Synonym: 2,6-di-tert-butyl-4-methylphenol,butylated hydroxytoluene,butylhydroxytoluene,2,6-di-tert-butyl-p-cresol,2,6-di-t-butyl-4-methylphenol,ionol,dbpc,dibunol,stavox,bht PubChem CID: 31404 ChEBI: CHEBI:34247 IUPAC Name: 2,6-ditert-butyl-4-methylphenol SMILES: CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C