man

Test

CAS RN 128446-35-5

OOHOHOOHOHOOHOOOHHOHOOOHOHOHOOOHOOOHOHOHOOHHOOOHOOHOHOHOHHHHHHHOH

CAS RN 128446-35-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 1180.05
Molecular Formula: C44H75O36
InChi Key:
SMILES: OCC-*.[H][C@@]12O[C@H](CO)[C@@H](O[C@@]3([H])O[C@H](CO)[C@@H](O[C@@]4([H])O[C@H](CO)[C@@H](O[C@@]5([H])O[C@H](CO)[C@@H](O[C@@]6([H])O[C@H](CO)[C@@H](O[C@@]7([H])O[C@H](CO)[C@@H](O[C@@]8([H])O[C@H](CO)[C@@H](O1)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)[C@H](O)[C@H]2O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)