CAS RN 1287-13-4
CAS RN 1287-13-4
IUPAC Name:
ruthenocene
Synonyms:
ruthenocene
Molecular Weight (g/mol):
226.22
Molecular Formula:
C10H5Ru
InChi Key:
—
SMILES:
c1cc[c-]c1.[C-]12[C-]3[C-]4[C-]5[C-]1[Ru]2345
1
–
1
of
1
results
1
Bis(cyclopentadienyl)ruthenium, Ru 43.2% min, Thermo Scientific Chemicals
CAS: 1287-13-4 MDL Number: MFCD00001442 Synonym: Ruthenocene