man

Test

CAS RN 130-40-5

H3CCH3NNONHONOPOOHHOOHOHOHNa

CAS RN 130-40-5

IUPAC Name: [(5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl)oxy]phosphonic acid sodium
Synonyms: riboflavin monophosphate sodium
Molecular Weight (g/mol): 479.34
Molecular Formula: C17H21N4NaO9P
InChi Key: WXEASYCOEACRIK-UHFFFAOYNA-N
SMILES: [Na].CC1=C(C)C=C2N(CC(O)C(O)C(O)COP(O)(O)=O)C3=NC(=O)NC(=O)C3=NC2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results