CAS RN 130018-87-0
CAS RN 130018-87-0
IUPAC Name:
2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dihydrochloride
Synonyms:
cetirizine dihydrochloride
Molecular Weight (g/mol):
461.81
Molecular Formula:
C21H27Cl3N2O3
InChi Key:
PGLIUCLTXOYQMV-UHFFFAOYNA-N
SMILES:
Cl.Cl.OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Levocetirizine dihydrochloride
CAS: 130018-87-0 Molecular Formula: C21H25ClN2O3·2HCl