CAS RN 1303-86-2
CAS RN 1303-86-2
Boron oxide, 98%, Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo(oxoboranyloxy)borane SMILES: O=BOB=O
Boron oxide, 98%, -40 mesh, Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo[(oxoboranyl)oxy]borane SMILES: O=BOB=O
Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo(oxoboranyloxy)borane SMILES: O=BOB=O
Boron oxide, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo[(oxoboranyl)oxy]borane SMILES: O=BOB=O
Boron oxide, 99.98% (metals basis), Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo[(oxoboranyl)oxy]borane SMILES: O=BOB=O
Boron oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific™
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 IUPAC Name: oxo(oxoboranyloxy)borane SMILES: O=BOB=O
Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 SMILES: O=BOB=O
Boron oxide, 97.5% min, typically 98.5%, Thermo Scientific Chemicals
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 SMILES: O=BOB=O
Boric Anhydride, Purified, J.T. Baker™
CAS: 1303-86-2 Molecular Formula: B2O3 Molecular Weight (g/mol): 69.62 MDL Number: MFCD00011315 InChI Key: JKWMSGQKBLHBQQ-UHFFFAOYSA-N Synonym: boron trioxide,boric oxide,boric anhydride,boric acid anhydride,oxo oxoboranyloxy borane,boronoxide,diboron trioxide,boron sesquioxide,boron oxide,boron oxide 500g PubChem CID: 518682 ChEBI: CHEBI:30163 SMILES: O=BOB=O