CAS RN 1303-96-4
CAS RN 1303-96-4
IUPAC Name:
disodium decahydrate oxoboranyl ({oxido[(oxoboranyl)oxy]boranyl}oxy)boronate
Synonyms:
borax
Molecular Weight (g/mol):
381.36
Molecular Formula:
B4H20Na2O17
InChi Key:
OJPFLODMGUZMKD-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B(OB=O)OB([O-])OB=O
1
–
15
of
23
results
1
Sodium tetraborate decahydrate, 99+%, Thermo Scientific Chemicals
CAS: 1303-96-4 Molecular Formula: B4Na2O7·10H2O MDL Number: MFCD00149193 Synonym: Borax; Sodium borate (tetra) decahydrate
![EDGE](/content/dam/fishersci/glyphs/edge.png)
2
Sodium tetraborate decahydrate, 99.5+%, ACS reagent, Thermo Scientific Chemicals
CAS: 1303-96-4 Molecular Formula: B4Na2O7·10H2O Molecular Weight (g/mol): 381.36 MDL Number: MFCD00149193 Synonym: Borax,Sodium borate decahydrate
3
Di-Sodium Tetraborate Decahydrate, Certified AR for Analysis, Fisher Chemical™
CAS: 1303-96-4 Molecular Formula: B4Na2O7·10H2O MDL Number: 149193
4
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
CAS: 1303-96-4 Molecular Formula: B4Na2O7·10H2O Molecular Weight (g/mol): 381.36 MDL Number: MFCD00149193 Synonym: Borax,Sodium borate decahydrate
12
Sodium tetraborate decahydrate, Honeywell Fluka™
CAS: 1303-96-4 MDL Number: MFCD00149193
13
Sodium Tetraborate Decahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
CAS: 1303-96-4 MDL Number: MFCD00149193
15
Standard Buffer Borate, pH 9.225, NIST, Chem-Lab
CAS: 1303-96-4 Molecular Formula: Na2B4O7•10H2O
- 1
- 2