man

Test

CAS RN 13116-27-3

HNNH2I

CAS RN 13116-27-3

IUPAC Name: (4-iodophenyl)hydrazine
Synonyms: (4-iodophenyl)hydrazine
Molecular Weight (g/mol): 234.04
Molecular Formula: C6H7IN2
InChi Key: IQMLCMKMSBMMGR-UHFFFAOYSA-N
SMILES: NNC1=CC=C(I)C=C1
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results