man

Test

CAS RN 1312-76-1

KOHSiOO

CAS RN 1312-76-1

IUPAC Name: potassium hydroxysilanoylolate
Synonyms: potassium silicate
Molecular Weight (g/mol): 116.19
Molecular Formula: HKO3Si
InChi Key: WMWBFVSTSRDWRL-UHFFFAOYSA-N
SMILES: [K+].O[Si]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results