man

Test

CAS RN 13122-90-2

H3CCH3CH3OONHOHNOHO

CAS RN 13122-90-2

IUPAC Name: 2-(2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanamido)-3-phenylpropanoic acid
Synonyms: 2-{2-[(tert-butoxycarbonyl)amino]-3-phenylpropanamido}-3-phenylpropanoic acid
Molecular Weight (g/mol): 412.49
Molecular Formula: C23H28N2O5
InChi Key: NNOBHAOOLCEJBL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)NC(CC1=CC=CC=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)