man

Test

CAS RN 13126-12-0

RbONOO

CAS RN 13126-12-0

IUPAC Name: rubidium(1+) nitrate
Synonyms: rubidium nitrate
Molecular Weight (g/mol): 147.47
Molecular Formula: NO3Rb
InChi Key: RTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES: [Rb+].[O-][N+]([O-])=O
Percent Purity 
  • (2)
  • (5)
  • (3)
Physical Form 
  • (8)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (3)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (8)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)