man

Test

CAS RN 13134-76-4

H3COHOSO

CAS RN 13134-76-4

IUPAC Name: (2Z)-2-[hydroxy(methoxy)methylidene]-2,3-dihydro-1-benzothiophen-3-one
Synonyms: (2Z)-2-[hydroxy(methoxy)methylidene]-1-benzothiophen-3-one
Molecular Weight (g/mol): 208.23
Molecular Formula: C10H8O3S
InChi Key: JVUOTPTULVZUTL-KTKRTIGZSA-N
SMILES: CO\C(O)=C1/SC2=CC=CC=C2C1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results