man

Test

CAS RN 1314-32-5

OTlOTlO

CAS RN 1314-32-5

IUPAC Name: oxo[(oxothallanyl)oxy]thallane
Synonyms: thallium(III) oxide
Molecular Weight (g/mol): 456.76
Molecular Formula: O3Tl2
InChi Key: QTQRFJQXXUPYDI-UHFFFAOYSA-N
SMILES: O=[Tl]O[Tl]=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results