man

Test

CAS RN 1315-09-9

ZnSe

CAS RN 1315-09-9

IUPAC Name: zinc selandiide
Synonyms: zinc selenide
Molecular Weight (g/mol): 144.35
Molecular Formula: SeZn
InChi Key: ALPUSWLPFCNUJH-UHFFFAOYSA-N
SMILES: [Zn].[Se--]
Quantity 
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (2)
  • (5)
Physical Form 
  • (2)
  • (2)
  • (5)
  • (2)
Grade 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)