man

Test

CAS RN 1317-41-5

CuSe

CAS RN 1317-41-5

IUPAC Name: copper(2+) selandiide
Synonyms: copper(2+) selandiide
Molecular Weight (g/mol): 142.52
Molecular Formula: CuSe
InChi Key: QMULOZLYOQCZOH-UHFFFAOYSA-N
SMILES: [Cu++].[Se--]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results