man

Test

CAS RN 13195-50-1

ONOSBr

CAS RN 13195-50-1

IUPAC Name: 2-bromo-5-nitrothiophene
Synonyms: 2-bromo-5-nitrothiophene
Molecular Weight (g/mol): 208.03
Molecular Formula: C4H2BrNO2S
InChi Key: ZPNFMDYBAQDFDY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)S1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)