man

Test

CAS RN 132-65-0

S

CAS RN 132-65-0

IUPAC Name: 8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: dibenzothiophene diphenylene sulfide dibenzo(b,d)thiophene 2,2'-biphenylylene sulfide
Molecular Weight (g/mol): 184.26
Molecular Formula: C12H8S
InChi Key: IYYZUPMFVPLQIF-UHFFFAOYSA-N
SMILES: S1C2=C(C=CC=C2)C2=C1C=CC=C2
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (6)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (4)
  • (1)