man

Test

CAS RN 1323966-36-4

CH3FFFFNH2

CAS RN 1323966-36-4

IUPAC Name: 1-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 207.17
Molecular Formula: C9H9F4N
InChi Key: RVXWFCFVRIODHK-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=C(F)C=C1C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results