man

Test

CAS RN 1323966-38-6

FOFFFOCl

CAS RN 1323966-38-6

IUPAC Name: 4-fluoro-2-(trifluoromethoxy)benzoyl chloride
Synonyms: 4-fluoro-2-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 242.55
Molecular Formula: C8H3ClF4O2
InChi Key: RAPKOVZLHNOHEJ-UHFFFAOYSA-N
SMILES: FC1=CC(OC(F)(F)F)=C(C=C1)C(Cl)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results