man

Test

CAS RN 1323966-42-2

CH3OFOClCl

CAS RN 1323966-42-2

IUPAC Name: 4-chloro-3-ethoxy-2-fluorobenzoyl chloride
Synonyms: 4-chloro-3-ethoxy-2-fluorobenzoyl chloride
Molecular Weight (g/mol): 237.05
Molecular Formula: C9H7Cl2FO2
InChi Key: XTWYKHZMPKQVSM-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(C(Cl)=O)=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results