CAS RN 1326283-60-6
CAS RN 1326283-60-6
IUPAC Name:
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms:
5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol):
267.11
Molecular Formula:
C13H19BFNO3
InChi Key:
JWACNMHKJHEWAQ-UHFFFAOYSA-N
SMILES:
COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results
1
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
CAS: 1326283-60-6 Molecular Formula: C13H19BFNO3 Molecular Weight (g/mol): 267.11 MDL Number: MFCD22419265 InChI Key: JWACNMHKJHEWAQ-UHFFFAOYSA-N Synonym: 4-amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester,5-fluoro-2-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline PubChem CID: 67363295 IUPAC Name: 5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline SMILES: COC1=C(N)C=C(F)C(=C1)B1OC(C)(C)C(C)(C)O1