CAS RN 132706-25-3
CAS RN 132706-25-3
IUPAC Name:
3-phenoxythiophene-2-carbaldehyde
Synonyms:
3-phenoxythiophene-2-carbaldehyde
Molecular Weight (g/mol):
204.24
Molecular Formula:
C11H8O2S
InChi Key:
QHJDRZXMSDASSH-UHFFFAOYSA-N
SMILES:
O=CC1=C(OC2=CC=CC=C2)C=CS1
1
–
1
of
1
results
1
3-Phenoxythiophene-2-carbaldehyde, 97%, Thermo Scientific™
CAS: 132706-25-3 Molecular Formula: C11H8O2S Molecular Weight (g/mol): 204.24 MDL Number: MFCD00178748 InChI Key: QHJDRZXMSDASSH-UHFFFAOYSA-N Synonym: 2-thiophenecarboxaldehyde,3-phenoxy,3-phenoxythiophene-2-carboxaldehyde,acmc-1c0te,3-phenoxy-2-thiophenecarbaldehyde,3-phenoxythiophen-2-carboxaldehyde,3-phenoxy-2-thiophenecarboxaldehyde PubChem CID: 2778987 IUPAC Name: 3-phenoxythiophene-2-carbaldehyde SMILES: O=CC1=C(OC2=CC=CC=C2)C=CS1