man

Test

CAS RN 132740-43-3

FNCO

CAS RN 132740-43-3

IUPAC Name: 1-fluoro-4-(isocyanatomethyl)benzene
Synonyms: 1-fluoro-4-(isocyanatomethyl)benzene
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: HHSIWJYERNCLKQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(CN=C=O)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results