man

Test

CAS RN 132746-60-2

CH3OHNN

CAS RN 132746-60-2

IUPAC Name: (2S,3S)-2-(diphenylmethyl)-N-[(2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine
Synonyms: (2S,3S)-2-(diphenylmethyl)-N-[(2-methoxyphenyl)methyl]-1-azabicyclo[2.2.2]octan-3-amine
Molecular Weight (g/mol): 412.58
Molecular Formula: C28H32N2O
InChi Key: FLNYLINBEZROPL-NSOVKSMOSA-N
SMILES: COC1=CC=CC=C1CN[C@H]1C2CCN(CC2)[C@H]1C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results