man

Test

CAS RN 133-37-9

OHOHHOOOHO

CAS RN 133-37-9

IUPAC Name: 2,3-dihydroxybutanedioic acid
Synonyms: (.+-.)-tartaric acid meso-TARTARIC ACID
Molecular Weight (g/mol): 150.09
Molecular Formula: C4H6O6
InChi Key: FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES: OC(C(O)C(O)=O)C(O)=O
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)