CAS RN 1336-21-6
CAS RN 1336-21-6
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Buffertlösning pH 10,00 (+/-0,024 @ 25oC) Färgad blå Specpure NIST Spårbar, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Ammoniaklösning, 35 %, certifierad AR för analys, d=0,88, Fisher Chemical™
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 LEDER: N.O
Ammoniaklösning, 35 %, extra ren, SLR, d=0,88, Fisher Chemical™
CAS: 1336-21-6 Molekylformel: H5NO MDL-nummer: 11418
Ammoniaklösning, 25 %, certifierad AR för analys, d=0,91, Fisher Chemical™
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N IUPAC-namn: aminhydrat LEDER: N.O
Ammonium hydroxide, for analysis, 28-30 wt% solution of NH3 in water
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 LEDER: N.O
Ammoniumhydroxid, ACS-reagens, 28-30% lösning i vatten, Thermo Scientific Chemicals
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 LEDER: N.O
Ammoniumhydroxid, för analys, 25 % fri ammoniak, lösning i vatten, Thermo Scientific Chemicals
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 LEDER: N.O
Ammonium hydroxide, extra pure, 25% solution in water
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC-namn: aminhydrat LEDER: N.O
Buffertlösning pH 7,00 (+/-0,024 @ 25oC) Ingen Färg Specpure NIST Spårbar, Thermo Scientific Chemicals
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Ammoniaklösning, 20 %, certifierad AR för analys, d=0,92, Fisher Chemical™
CAS: 1336-21-6 Molekylformel: H5NO Molekylvikt (g/mol): 35.05 MDL-nummer: MFCD00066650 InChI-nyckel: VHUUQVKOLVNVRT-UHFFFAOYSA-N IUPAC-namn: aminhydrat LEDER: N.O
Buffer solution, pH 12.00 (+/-0.01 @ 25oC), No Color, Specpure, NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol