man

Test

CAS RN 134-20-3

CH3OOH2N

CAS RN 134-20-3

IUPAC Name: methyl 2-aminobenzoate
Synonyms: methyl anthranilate
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: VAMXMNNIEUEQDV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N
Percent Purity 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)