man

Test

CAS RN 13428-80-3

CH3OH3COH3COH3COHf

CAS RN 13428-80-3

IUPAC Name: hafnium(4+) tetraethanolate
Synonyms: hafnium(4+) tetrakis(ethoxide)
Molecular Weight (g/mol): 358.73
Molecular Formula: C8H20HfO4
InChi Key: BFIMXCBKRLYJQO-UHFFFAOYSA-N
SMILES: [Hf+4].CC[O-].CC[O-].CC[O-].CC[O-]
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)