CAS RN 134448-10-5
CAS RN 134448-10-5
IUPAC Name:
(2S)-1-[(2S,3S)-3-methyl-2-{[(3S)-3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl]pyrrolidine-2-carboxylic acid
Synonyms:
(2S)-1-[(2S,3S)-3-methyl-2-{[(3S)-3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol):
383.45
Molecular Formula:
C18H29N3O6
InChi Key:
ZEZGJKSEBRELAS-KSFNBINOSA-N
SMILES:
CCCNC(=O)[C@H]1OC1C(=O)N[C@@H]([C@@H](C)CC)C(=O)N1CCC[C@H]1C(O)=O
1
–
2
of
2
results