man

Test

CAS RN 1345-04-6

SbSbSSS

CAS RN 1345-04-6

IUPAC Name: antimony sulfane
Synonyms: antimony trisulfide
Molecular Weight (g/mol): 155.84
Molecular Formula: H2SSb
InChi Key: OLYFKZHUYAZYQW-UHFFFAOYSA-N
SMILES: S.[Sb]
Physical Form 
  • (2)
  • (6)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (6)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)