man

Test

CAS RN 13455-24-8

KOHIOOOIOO

CAS RN 13455-24-8

IUPAC Name: potassium iodic acid iodate
Synonyms: potassium iodic acid iodate
Molecular Weight (g/mol): 389.91
Molecular Formula: HI2KO6
InChi Key: ACAYDTMSDROWHW-UHFFFAOYSA-M
SMILES: [K+].O[I](=O)=O.[O-][I](=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)