man

Test

CAS RN 13472-33-8

OReOOONa

CAS RN 13472-33-8

IUPAC Name: sodium rheniumoylolate
Synonyms: sodium perrhenate
Molecular Weight (g/mol): 273.19
Molecular Formula: NaO4Re
InChi Key: KMISVFTVBOPTAI-UHFFFAOYSA-N
SMILES: [Na+].[O-][Re](=O)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Physical Form 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)