CAS RN 135236-72-5
CAS RN 135236-72-5
IUPAC Name:
3-hydroxy-3-methylbutanoate
Synonyms:
β-hydroxyisovalerate
Molecular Weight (g/mol):
117.13
Molecular Formula:
C5H9O3
InChi Key:
AXFYFNCPONWUHW-UHFFFAOYSA-M
SMILES:
CC(C)(O)CC([O-])=O
1
–
1
of
1
results
1
Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%, Thermo Scientific Chemicals
CAS: 135236-72-5 Molecular Formula: C10H18CaO6 Molecular Weight (g/mol): 274.33 MDL Number: MFCD01318562 InChI Key: WLJUMPWVUPNXMF-UHFFFAOYSA-L Synonym: calcium 3-hydroxy-3-methyl butyrate PubChem CID: 131847069 SMILES: [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O