man

Test

CAS RN 13536-59-9

1HBr

CAS RN 13536-59-9

IUPAC Name: proton bromide
Synonyms: proton bromide
Molecular Weight (g/mol): 80.91
Molecular Formula: BrH
InChi Key: CPELXLSAUQHCOX-FTGQXOHASA-N
SMILES: [1H+].[Br-]

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results