man

Test

CAS RN 135654-16-9

ClOH2N

CAS RN 135654-16-9

IUPAC Name: 3-(chloromethyl)benzamide
Synonyms: 3-(chloromethyl)benzamide
Molecular Weight (g/mol): 169.61
Molecular Formula: C8H8ClNO
InChi Key: OOYATKQQGVPYQR-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(CCl)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results