CAS RN 135748-34-4
CAS RN 135748-34-4
IUPAC Name:
2-chloro-4,5-difluorobenzonitrile
Synonyms:
2-chloro-4,5-difluorobenzonitrile
Molecular Weight (g/mol):
173.55
Molecular Formula:
C7H2ClF2N
InChi Key:
AKZDICFDUJUQRX-UHFFFAOYSA-N
SMILES:
FC1=CC(Cl)=C(C=C1F)C#N
1
–
1
of
1
results
1
2-Chloro-4,5-difluorobenzonitrile, 97%, Thermo Scientific™
CAS: 135748-34-4 Molecular Formula: C7H2ClF2N Molecular Weight (g/mol): 173.55 MDL Number: MFCD02871619 InChI Key: AKZDICFDUJUQRX-UHFFFAOYSA-N PubChem CID: 4085849 IUPAC Name: 2-chloro-4,5-difluorobenzonitrile SMILES: FC1=CC(Cl)=C(C=C1F)C#N