man

Test

CAS RN 13608-87-2

H3COClClCl

CAS RN 13608-87-2

IUPAC Name: 1-(2,3,4-trichlorophenyl)ethan-1-one
Synonyms: 1-(2,3,4-trichlorophenyl)ethanone
Molecular Weight (g/mol): 223.48
Molecular Formula: C8H5Cl3O
InChi Key: BXJZZJYNVIDEKG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(Cl)=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results