man

Test

CAS RN 13610-02-1

CHO

CAS RN 13610-02-1

IUPAC Name: (prop-2-yn-1-yloxy)benzene
Synonyms: (prop-2-yn-1-yloxy)benzene
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: AIQRJSXKXVZCJO-UHFFFAOYSA-N
SMILES: C#CCOC1=CC=CC=C1
Boiling Point 
  • (7)
  • (2)
Percent Purity 
  • (3)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (7)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)