CAS RN 13675-18-8
CAS RN 13675-18-8
IUPAC Name:
(dihydroxyboranyl)boronic acid
Synonyms:
tetrahydroxydiborane diborane(4)tetrol hypodiboric acid
Molecular Weight (g/mol):
89.65
Molecular Formula:
B2H4O4
InChi Key:
SKOWZLGOFVSKLB-UHFFFAOYSA-N
SMILES:
OB(O)B(O)O
1
–
1
of
1
results
1
Tetrahydroxydiboron, 95%, Thermo Scientific Chemicals
CAS: 13675-18-8 Molecular Formula: B2H4O4 Molecular Weight (g/mol): 89.65 MDL Number: MFCD05663888 InChI Key: SKOWZLGOFVSKLB-UHFFFAOYSA-N IUPAC Name: (dihydroxyboranyl)boronic acid SMILES: OB(O)B(O)O
Promotions are available