CAS RN 13720-94-0
CAS RN 13720-94-0
IUPAC Name:
ethyl 4-chloroquinoline-3-carboxylate
Synonyms:
ethyl 4-chloroquinoline-3-carboxylate
Molecular Weight (g/mol):
235.67
Molecular Formula:
C12H10ClNO2
InChi Key:
DWXQUAHMZWZXHP-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CN=C2C=CC=CC2=C1Cl
1
–
1
of
1
results
1
Ethyl 4-chloroquinoline-3-carboxylate, 96%, Thermo Scientific™
CAS: 13720-94-0 Molecular Formula: C12H10ClNO2 Molecular Weight (g/mol): 235.667 MDL Number: MFCD00173407 InChI Key: DWXQUAHMZWZXHP-UHFFFAOYSA-N Synonym: ethyl 4-chloro-3-quinolinecarboxylate,4-chloroquinoline-3-carboxylic acid ethyl ester,ethyl 4-chloro-quinoline-3-carboxylate,3-quinolinecarboxylic acid, 4-chloro-, ethyl ester,pubchem19592,acmc-20ai5d,ethylchloroquinolinecarboxylate,ethyl 4-chloroquinolin-3-carboxylate,ethyl 4-chloro-3-quinoline carboxylate,ethyl 4-chloranylquinoline-3-carboxylate PubChem CID: 268963 IUPAC Name: ethyl 4-chloroquinoline-3-carboxylate SMILES: CCOC(=O)C1=C(C2=CC=CC=C2N=C1)Cl