man

Test

CAS RN 1373920-74-1

H3COFOClF

CAS RN 1373920-74-1

IUPAC Name: 4-butoxy-2,6-difluorobenzoyl chloride
Synonyms: 4-butoxy-2,6-difluorobenzoyl chloride
Molecular Weight (g/mol): 248.65
Molecular Formula: C11H11ClF2O2
InChi Key: LCYFGUHGFICGBL-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(C(Cl)=O)C(F)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results