man

Test

CAS RN 1373920-89-8

CH3OFNCl

CAS RN 1373920-89-8

IUPAC Name: 4-chloro-3-ethoxy-2-fluorobenzonitrile
Synonyms: 4-chloro-3-ethoxy-2-fluorobenzonitrile
Molecular Weight (g/mol): 199.61
Molecular Formula: C9H7ClFNO
InChi Key: SVYHZDFUKRNQNH-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(C#N)=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results