CAS RN 1373920-97-8
CAS RN 1373920-97-8
IUPAC Name:
6-fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
Synonyms:
6-fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
206.14
Molecular Formula:
C9H6F4O
InChi Key:
CLGGHNMZJHOQOY-UHFFFAOYSA-N
SMILES:
CC1=C(C(C=O)=C(F)C=C1)C(F)(F)F
1
–
1
of
1
results
1
6-Fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
CAS: 1373920-97-8 Molecular Formula: C9H6F4O Molecular Weight (g/mol): 206.14 MDL Number: MFCD22201035 InChI Key: CLGGHNMZJHOQOY-UHFFFAOYSA-N Synonym: 6-fluoro-3-methyl-2-trifluoromethyl benzaldehyde PubChem CID: 86277637 IUPAC Name: 6-fluoro-3-methyl-2-(trifluoromethyl)benzaldehyde SMILES: CC1=C(C(=C(C=C1)F)C=O)C(F)(F)F