CAS RN 13749-38-7
CAS RN 13749-38-7
IUPAC Name:
1,2-dibromo-1,1,2-trichloroethane
Synonyms:
1,2-dibromo-1,1,2-trichloroethane
Molecular Weight (g/mol):
291.19
Molecular Formula:
C2HBr2Cl3
InChi Key:
VXKOVUFOVCJHBA-UHFFFAOYNA-N
SMILES:
ClC(Br)C(Cl)(Cl)Br
1
–
1
of
1
results
1
1,2-Dibromo-1,1,2-trichloroethane, 99%, Thermo Scientific™
CAS: 13749-38-7 Molecular Formula: C2HBr2Cl3 Molecular Weight (g/mol): 291.2 MDL Number: MFCD00018841 InChI Key: VXKOVUFOVCJHBA-UHFFFAOYSA-N Synonym: ethane,1,2-dibromo-1,1,2-trichloro,acmc-20ap6p,1,2-dibromotrichloro ethane PubChem CID: 114562 IUPAC Name: 1,2-dibromo-1,1,2-trichloroethane SMILES: C(C(Cl)(Cl)Br)(Cl)Br