man

Test

CAS RN 137504-86-0

HOBOHClF

CAS RN 137504-86-0

IUPAC Name: (4-chloro-3-fluorophenyl)boronic acid
Synonyms: 4-chloro-3-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: CMJQIHGBUKZEHP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C(F)=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results