CAS RN 13765-74-7
CAS RN 13765-74-7
IUPAC Name:
disilver(1+) dioxomolybdenumbis(olate)
Synonyms:
silver molybdate
Molecular Weight (g/mol):
375.68
Molecular Formula:
Ag2MoO4
InChi Key:
MHLYOTJKDAAHGI-UHFFFAOYSA-N
SMILES:
[Ag+].[Ag+].[O-][Mo]([O-])(=O)=O
1
–
1
of
1
results
1
Silver molybdenum oxide, 99%, Thermo Scientific Chemicals
CAS: 13765-74-7 MDL Number: MFCD00053384 Synonym: Silver(I) molybdate