CAS RN 13767-32-3
CAS RN 13767-32-3
IUPAC Name:
zinc(2+) molybdenum tetraoxidandiide
Synonyms:
zinc molybdate
Molecular Weight (g/mol):
225.33
Molecular Formula:
MoO4Zn
InChi Key:
CEKVBJSJDXQDQR-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[O--].[Zn++].[Mo]
1
–
1
of
1
results
1
Zinc molybdenum oxide, 98%, Thermo Scientific Chemicals
CAS: 13767-32-3 Molecular Formula: ZnMoO4 MDL Number: MFCD00049622 Synonym: Zinc molybdate