man

Test

CAS RN 13790-39-1

CH3ONNClOCH3

CAS RN 13790-39-1

IUPAC Name: 4-chloro-6,7-dimethoxyquinazoline
Synonyms: 4-chloro-6,7-dimethoxyquinazoline
Molecular Weight (g/mol): 224.64
Molecular Formula: C10H9ClN2O2
InChi Key: LLLHRNQLGUOJHP-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(Cl)=NC=NC2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results