man

Test

CAS RN 138-15-8

HClNH2HOOOHO

CAS RN 138-15-8

IUPAC Name: hydrogen 2-aminopentanedioic acid chloride
Synonyms: hydrogen glutaminsaeure chloride
Molecular Weight (g/mol): 183.59
Molecular Formula: C5H10ClNO4
InChi Key: RPAJSBKBKSSMLJ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC(CCC(O)=O)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)